4-Fluoro-3-methoxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester

A297915-1G A297915-250MG A297915-5G
76827-800EA 62.6 USD
76827-800 76827-802 76827-804
4-Fluoro-3-methoxyphenylboronic acid pinacol ester 98%
4-Fluoro-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
MDL Number: MFCD11227055
CAS Number: 425378-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR