7-Fluoro-1H-indole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A297050-100MG A297050-1G A297050-250MG
76827-782EA 35.01 USD
76827-782 76827-784 76827-786
7-Fluoro-1H-indole-3-carboxylic acid 98%
7-Fluoro-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.148 g/mol
MDL Number: MFCD09954940
CAS Number: 858515-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR