2-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A296314-100MG A296314-1G A296314-250MG A296314-5G
76827-770EA 31 USD
76827-770 76827-772 76827-774 76827-776
2-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%
2-Chloro-4-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MDL Number: MFCD11040272
CAS Number: 345226-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR