4-Isopropylphenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A294349-1G A294349-250MG A294349-25G A294349-5G
76827-750EA 28 USD
76827-750 76827-752 76827-754 76827-756
4-Isopropylphenylboronic acid pinacol ester 96%
4-Isopropylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₂
MW: 246.1529 g/mol
MDL Number: MFCD15143592
CAS Number: 325142-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR