Methyl 2-ethyl-3-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A293520-100MG A293520-250MG
76827-746EA 101 USD
76827-746 76827-748
Methyl 2-ethyl-3-hydroxybenzoate 98%
Methyl 2-ethyl-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD22490933
CAS Number: 183108-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR