2-Amino-5-cyanobenzoic acid 97%

Supplier: Ambeed
Warning

A291422-1G A291422-250MG A291422-5G
76827-722EA 55 USD
76827-722 76827-724 77885-765
2-Amino-5-cyanobenzoic acid 97%
2-Amino-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD20485816
CAS Number: 99767-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR