3-Bromobenzanilide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-N-phenylbenzamide

A291002-10G A291002-250MG A291002-25G A291002-5G
76827-714EA 84.88 USD
76827-714 76827-716 76827-718 76827-720
3-Bromobenzanilide 95%
3-Bromobenzanilide

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
MW: 276.1300 g/mol
MDL Number: MFCD00087947
CAS Number: 63710-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR