(3R,4R)-rel-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A290444-10G A290444-25G
76827-696EA 269.47 USD
76827-696 76827-698
(3R,4R)-rel-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate 95%
(3R,4R)-rel-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.25 g/mol
MDL Number: MFCD06858457
CAS Number: 148214-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR