4-Bromo-2-methoxybenzotrifluoride 98%

Supplier: Ambeed
Warning

A290416-1G A290416-25G A290416-5G
76827-690EA 21.99 USD
76827-690 76827-692 76827-694
4-Bromo-2-methoxybenzotrifluoride 98%
4-Bromo-2-methoxybenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MDL Number: MFCD13194486
CAS Number: 944805-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR