4-Iodo-1H-pyrazolo[3,4-b]pyridine 98%

Supplier: Ambeed
Warning

A290011-100MG A290011-1G A290011-250MG
76827-682EA 7 USD
76827-682 76827-684 76827-686
4-Iodo-1H-pyrazolo[3,4-b]pyridine 98%
4-Iodo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
MDL Number: MFCD11846314
CAS Number: 861881-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR