4-Fluoro-1H-indole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A289855-250MG A289855-5G A289855-1G
76827-678EA 103.97 USD
76827-678 76827-680 77683-867
4-Fluoro-1H-indole-6-carboxylic acid 95%
4-Fluoro-1H-indole-6-carboxylic acid

Precaución: For research use only.

Formula: C₉H₆FNO₂
MW: 179.148 g/mol
MDL Number: MFCD07781705
CAS Number: 885520-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR