4-Chloro-6-fluoro-1H-indazole 95%

Supplier: Ambeed
Warning

A288728-100MG A288728-1G A288728-250MG
76827-670EA 37.14 USD
76827-670 76827-672 76827-674
4-Chloro-6-fluoro-1H-indazole 95%
4-Chloro-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
MW: 170.57 g/mol
MDL Number: MFCD07781447
CAS Number: 885520-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR