6-Bromo-2,4-dihydrospiro[benzo[b][1,4]dioxepine-3,1'-cyclopropane] 98%

Supplier: Ambeed
Warning

A287849-100MG A287849-1G A287849-250MG
76827-658EA 36.08 USD
76827-658 76827-660 76827-662
6-Bromo-2,4-dihydrospiro[benzo[b][1,4]dioxepine-3,1'-cyclopropane] 98%
6-Bromo-2,4-dihydrospiro[benzo[b][1,4]dioxepine-3,1'-cyclopropane]

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.11 g/mol
MDL Number: MFCD20231462
CAS Number: 1334499-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR