4-{[(4-Methoxybenzyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-(4-Methoxybenzyl)ureido)phenylboronic acid, pinacol ester, 1-(4-Methoxybenzyl)-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea

A286942-1G A286942-250MG A286942-5G
76827-636EA 73 USD
76827-636 76827-638 76827-640
4-{[(4-Methoxybenzyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%
4-{[(4-Methoxybenzyl)carbamoyl]amino}benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₂₇BN₂O₄
MDL Number: MFCD09027293
CAS Number: 874298-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR