(S)-5-(Benzyloxy)-2-(1,3-dioxoisoindolin-2-yl)-5-oxopentanoic acid 97%
Supplier: Ambeed
|
Warning
|
A286055-100MG
A286055-1G
A286055-250MG
A286055-5G
76827-620EA
21
USD
76827-620
76827-622
76827-624
76827-626
(S)-5-(Benzyloxy)-2-(1,3-dioxoisoindolin-2-yl)-5-oxopentanoic acid 97%
(S)-5-(Benzyloxy)-2-(1,3-dioxoisoindolin-2-yl)-5-oxopentanoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₁₇NO₆ |
MDL Number:
MFCD13152207 CAS Number: 88784-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...