1-Bromo-4-(bromomethyl)naphthalene 97%

Supplier: Ambeed
Danger

A284418-10G A284418-1G A284418-250MG A284418-25G A284418-5G
76827-606EA 230.22 USD
76827-606 76827-608 76827-610 76827-612 76827-614
1-Bromo-4-(bromomethyl)naphthalene 97%
1-Bromo-4-(bromomethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₈Br₂
MW: 299.9892 g/mol
MDL Number: MFCD07363811
CAS Number: 79996-99-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR