3-Bromo-2,4-difluoropyridine 95%

Supplier: Ambeed
Warning

A284104-1G A284104-250MG A284104-5G
76827-596EA 107 USD
76827-596 76827-598 77885-754
3-Bromo-2,4-difluoropyridine 95%
3-Bromo-2,4-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrF₂N
MDL Number: MFCD16609909
CAS Number: 1227502-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR