(S)-2-((S)-2-Cyclohexyl-2-(pyrazine-2-carboxamido)acetamido)-3,3-dimethylbutanoic acid 98+%

Supplier: Ambeed
Warning

A281911-1G A281911-250MG
76827-582EA 39.26 USD
76827-582 76827-584
(S)-2-((S)-2-Cyclohexyl-2-(pyrazine-2-carboxamido)acetamido)-3,3-dimethylbutanoic acid 98+%
(S)-2-((S)-2-Cyclohexyl-2-(pyrazine-2-carboxamido)acetamido)-3,3-dimethylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₄O₄
MW: 376.45 g/mol
MDL Number: MFCD11865138
CAS Number: 402958-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR