(6-Fluorobenzo[d]thiazol-2-yl)methanol 95%

Supplier: Ambeed
Warning

A281508-100MG A281508-250MG
76827-576EA 130.5 USD
76827-576 76827-578
(6-Fluorobenzo[d]thiazol-2-yl)methanol 95%
(6-Fluorobenzo[d]thiazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₆FNOS
MW: 183.20 g/mol
MDL Number: MFCD11977694
CAS Number: 197364-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR