4-(4-(4-Methylpiperazin-1-yl)piperidin-1-yl)aniline 95%

Supplier: Ambeed
Warning

A281464-100MG A281464-1G A281464-250MG A281464-5G
76827-568EA 20.16 USD
76827-568 76827-570 76827-572 76827-574
4-(4-(4-Methylpiperazin-1-yl)piperidin-1-yl)aniline 95%
4-(4-(4-Methylpiperazin-1-yl)piperidin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₆H₂₆N₄
MW: 274.40 g/mol
MDL Number: MFCD22493476
CAS Number: 959795-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR