4-Chloro-5-methyl-2-(methylthio)pyrimidine 97%

Supplier: Ambeed
Warning

A281338-10G A281338-250MG
76827-562EA 558.04 USD
76827-562 76827-564
4-Chloro-5-methyl-2-(methylthio)pyrimidine 97%
4-Chloro-5-methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂S
MW: 174.651 g/mol
MDL Number: MFCD09951961
CAS Number: 61044-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR