6-Chlorophenanthridine 95%

Supplier: Ambeed
Warning

A280556-10G A280556-1G A280556-250MG A280556-25G A280556-5G
76827-546EA 240 USD
76827-546 76827-548 76827-550 76827-552 76827-554
6-Chlorophenanthridine 95%
6-Chlorophenanthridine

Precaución: for Research Use Only

Formula: C₁₃H₈ClN
MDL Number: MFCD01628084
CAS Number: 15679-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR