5-Bromothiophene-2-carboxamide 96%

Supplier: Ambeed
Warning

A280485-100MG A280485-250MG A280485-1G A280485-5G
76827-542EA 19.1 USD
76827-542 76827-544 77593-736 77683-847
5-Bromothiophene-2-carboxamide 96%
5-Bromothiophene-2-carboxamide

Precaución: For research use only.

Formula: C₅H₄BrNOS
MDL Number: MFCD01922020
CAS Number: 76371-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR