(4-(1-((tert-Butoxycarbonyl)amino)cyclopropyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A279679-100MG A279679-1G A279679-250MG A279679-5G
76827-530EA 37 USD
76827-530 76827-532 76827-534 77885-747
(4-(1-((tert-Butoxycarbonyl)amino)cyclopropyl)phenyl)boronic acid 98%
(4-(1-((tert-Butoxycarbonyl)amino)cyclopropyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄
MDL Number: MFCD12546458
CAS Number: 1217500-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR