3-Chloro-2,4-difluorobromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-chloro-2,4-difluorobenzene

A279146-100G A279146-1G A279146-25G A279146-5G
76827-520EA 496.51 USD
76827-520 76827-522 76827-524 76827-526
3-Chloro-2,4-difluorobromobenzene 98%
3-Chloro-2,4-difluorobromobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
MDL Number: MFCD00142590
CAS Number: 201849-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR