Sodium bromoacetate 98%

Supplier: Ambeed
Danger

Synonyms: Sodium 2-bromoacetate

A278953-1G A278953-25G A278953-5G
76827-502EA 14 USD
76827-502 76827-504 76827-506
Sodium bromoacetate 98%
Sodium bromoacetate

Precaución: for Research Use Only

Formula: BrCH₂CO₂Na
MDL Number: MFCD01311832
CAS Number: 1068-52-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR