Pyridine-3,5-dicarboxamide 95%

Supplier: Ambeed
Warning

A278673-100MG A278673-1G A278673-250MG
76827-482EA 50 USD
76827-482 76827-484 76827-486
Pyridine-3,5-dicarboxamide 95%
Pyridine-3,5-dicarboxamide

Precaución: for Research Use Only

Formula: C₇H₇N₃O₂
MDL Number: MFCD00229426
CAS Number: 4663-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR