N-Ethyl-2(or 4)-methylbenzenesulfonamide ≥98% (mixture of isomers)

Supplier: Ambeed
Warning

A278671-100G A278671-25G A278671-500G
76827-476EA 14.86 USD
76827-476 76827-478 76827-480
N-Ethyl-2(or 4)-methylbenzenesulfonamide ≥98% (mixture of isomers)
N-Ethyl-2(or 4)-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
MDL Number: MFCD00152499
CAS Number: 8047-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR