4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester, 4-Boc-phenylboronic acid Pinacol ester, 2-(4-tert-Butoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)phenylboronic acid Pinacol ester

A277848-10G A277848-1G A277848-25G A277848-5G A277848-250MG
76827-440EA 30 USD
76827-440 76827-442 76827-444 76827-446 77885-740
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₄
MDL Number: MFCD03453055
CAS Number: 850568-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR