2-(Methylthio)-5,6,7,8-tetrahydroquinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

A276756-100MG A276756-1G A276756-250MG
76827-424EA 54.11 USD
76827-424 76827-426 76827-428
2-(Methylthio)-5,6,7,8-tetrahydroquinazolin-4(3H)-one 97%
2-(Methylthio)-5,6,7,8-tetrahydroquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₁₂N₂OS
MW: 196.2700 g/mol
MDL Number: MFCD08744204
CAS Number: 34170-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR