4-(Trifluoromethyl)-1H-indazole 97%

Supplier: Ambeed
Warning

A274046-1G A274046-250MG A274046-50MG A274046-5G
76827-398EA 134.74 USD
76827-398 76827-400 76827-402 76827-404
4-(Trifluoromethyl)-1H-indazole 97%
4-(Trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MDL Number: MFCD09263211
CAS Number: 1000339-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR