2-Fluoro-4-methoxynicotinic acid 95%

Supplier: Ambeed
Warning

A273937-100MG A273937-1G A273937-250MG A273937-5G
76827-392EA 47 USD
76827-392 76827-394 76827-396 77885-728
2-Fluoro-4-methoxynicotinic acid 95%
2-Fluoro-4-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD12828253
CAS Number: 1190315-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR