3-Chloro-5,6-dimethylpyrazin-2-amine 97%

Supplier: Ambeed
Warning

A272533-100MG A272533-1G A272533-250MG
76827-380EA 40.32 USD
76827-380 76827-382 76827-384
3-Chloro-5,6-dimethylpyrazin-2-amine 97%
3-Chloro-5,6-dimethylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
MDL Number: MFCD21234604
CAS Number: 39213-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR