Methyl 1,3-benzoxazole-6-carboxylate 98%

Supplier: Ambeed
Warning

A272117-100MG A272117-1G A272117-250MG
76827-352EA 15 USD
76827-352 76827-354 76827-356
Methyl 1,3-benzoxazole-6-carboxylate 98%
Methyl 1,3-benzoxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD14635902
CAS Number: 1305711-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR