4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl 95%

Supplier: Ambeed
Danger

A271553-1G A271553-5G A271553-100MG
76827-348EA 91 USD
76827-348 76827-350 77885-721
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl 95%
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
MDL Number: MFCD15146810
CAS Number: 147497-57-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR