1-(2-Bromophenyl)cyclopropane-1-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Carboxy-1-(2-bromophenyl)cyclopropane, 1-(2-Bromophenyl)cyclopropanecarboxylic acid, 1-(1-Carboxycycloprop-1-yl)-2-bromobenzene
A271076-1G
A271076-100MG
A271076-250MG
A271076-5G
76827-340EA
25
USD
76827-340
77885-714
77885-715
77885-716
1-(2-Bromophenyl)cyclopropane-1-carboxylic acid 97%
1-(2-Bromophenyl)cyclopropane-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉BrO₂ |
MDL Number:
MFCD07374434 CAS Number: 124276-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...