1-(Benzyloxy)-4-bromo-2-(trifluoromethyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzyloxy)-5-bromobenzotrifluoride

A268618-1G A268618-250MG
76827-280EA 31.83 USD
76827-280 76827-282
1-(Benzyloxy)-4-bromo-2-(trifluoromethyl)benzene 97%
1-(Benzyloxy)-4-bromo-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.13 g/mol
MDL Number: MFCD12913506
CAS Number: 169247-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR