3-(Dimethylamino)benzonitrile 97%

Supplier: Ambeed
Danger

A268208-1G A268208-250MG A268208-5G
76827-274EA 35 USD
76827-274 76827-276 76827-278
3-(Dimethylamino)benzonitrile 97%
3-(Dimethylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD00159008
CAS Number: 38803-30-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR