3-Hydroxytetrahydrothiophene 1,1-dioxide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrahydrothiophen-3-ol 1,1-dioxide, 3-Hydroxysulpholane
A267137-250MG
A267137-25G
A267137-5G
A267137-1G
76827-252EA
21.22
USD
76827-252
76827-254
76827-256
77683-819
3-Hydroxytetrahydrothiophene 1,1-dioxide 98%
3-Hydroxytetrahydrothiophene 1,1-dioxide
Precaución: For research use only.
Formula:
C₄H₈O₃S MW: 136.17 g/mol |
MDL Number:
MFCD00154867 CAS Number: 13031-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...