(R)-3,3'-Di(anthracenyl-9-yl)-[1,1'-binapthalene]-2,2'-diol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-3,3'-Di(anthracen-9-yl)-[1,1'-binaphthalene]-2,2'-diol
A266800-100MG
A266800-250MG
A266800-1G
76827-242EA
111.4
USD
76827-242
76827-244
77683-818
(R)-3,3'-Di(anthracenyl-9-yl)-[1,1'-binapthalene]-2,2'-diol 98%
(R)-3,3'-Di(anthracenyl-9-yl)-[1,1'-binapthalene]-2,2'-diol
Precaución: For research use only.
Formula:
C₄₈H₃₀O₂ MW: 638.751 g/mol |
MDL Number:
MFCD09027270 CAS Number: 361342-49-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...