(S)-1-(3-Chlorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A266791-1G A266791-250MG A266791-5G
76827-236EA 92.3 USD
76827-236 76827-238 76827-240
(S)-1-(3-Chlorophenyl)ethanol 95%
(S)-1-(3-Chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.60 g/mol
MDL Number: MFCD08458908
CAS Number: 135145-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR