4-(4-Bromophenethyl)morpholine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A265131-100MG A265131-250MG A265131-1G
76827-208EA 37.14 USD
76827-208 76827-210 77683-809
4-(4-Bromophenethyl)morpholine 95%, Ambeed.Inc
4-(4-Bromophenethyl)morpholine

Precaución: For research use only.

Formula: C₁₂H₁₆BrNO
MW: 270.17 g/mol
MDL Number: MFCD11855960
CAS Number: 736991-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR