2,4-Dibromo-6-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A265070-250MG A265070-5G
76827-204EA 29.71 USD
76827-204 77683-808
2,4-Dibromo-6-fluorobenzaldehyde 98%
2,4-Dibromo-6-fluorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃Br₂FO
MW: 281.905 g/mol
MDL Number: MFCD03094089
CAS Number: 205683-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR