CH5424802 (Alectinib) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Alectinib, 9-Ethyl-6,6-dimethyl-8-(4-morpholinopiperidin-1-yl)-11-oxo-6,11-dihydro-5H-benzo[b]carbazole-3-carbonitrile, 9-Ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[b]carbazol-3-carbonitrile
A264992-10MG
A264992-5MG
A264992-1G
A264992-250MG
A264992-100MG
A264992-5G
76827-196EA
6.37
USD
76827-196
76827-198
77612-538
77612-540
77683-806
77683-807
CH5424802 (Alectinib) 98%
CH5424802 (Alectinib)
Precaución: For research use only.
Formula:
C₃₀H₃₄N₄O₂ MW: 482.62 g/mol |
MDL Number:
MFCD19440988 CAS Number: 1256580-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...