4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one ≥95%

Supplier: Ambeed
Warning

Synonyms: 2'-O-(2-Methoxyethyl)-cytidine

A264907-100MG A264907-5G A264907-250MG
76827-192EA 39.26 USD
76827-192 76827-194 77683-805
4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one ≥95%
4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

Precaución: For research use only.

Formula: C₁₂H₁₉N₃O₆
MW: 301.29 g/mol
MDL Number: MFCD17215637
CAS Number: 223777-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR