Ethyl 2-(5-bromopyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A263861-1G A263861-250MG A263861-5G
76827-172EA 17 USD
76827-172 76827-174 77612-524
Ethyl 2-(5-bromopyridin-2-yl)acetate 95%
Ethyl 2-(5-bromopyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD10566270
CAS Number: 1060814-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR