2-Chloro-4-nitropyridin-3-ol 95%

Supplier: Ambeed
Warning

A263157-250MG A263157-1G A263157-5G
76827-156EA 43 USD
76827-156 77885-688 77885-689
2-Chloro-4-nitropyridin-3-ol 95%
2-Chloro-4-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MDL Number: MFCD11847780
CAS Number: 15128-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR