S-BAIBA 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (S)-b-aminoisobutyric acid, L-3-Amino-isobutyric acid, (S)-3-Amino-2-methylpropanoic acid

A262596-1G A262596-250MG A262596-100MG A262596-5G A262596-25G
76827-138EA 125 USD
76827-138 76827-140 77612-514 77612-516 77683-800
S-BAIBA 95%, Ambeed.Inc
S-BAIBA

Precaución: For research use only.

Formula: C₄H₉NO₂
MDL Number: MFCD07372875
CAS Number: 4249-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR