(3-Fluoropyridin-2-yl)methanamine hydrochloride 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A262557-5G A262557-250MG
76827-136EA 193 USD
76827-136 77683-799
(3-Fluoropyridin-2-yl)methanamine hydrochloride 98%, Ambeed.Inc
(3-Fluoropyridin-2-yl)methanamine hydrochloride

Precaución: For research use only.

Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD11505887
CAS Number: 1260903-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR