5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-1,3-thiazol-2-yl)methanol, (5-Bromothiazol-2-yl)methanol

A261957-100MG A261957-10G A261957-1G A261957-250MG A261957-5G
76827-126EA 49.87 USD
76827-126 76827-128 76827-130 76827-132 76827-134
5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%
5-Bromo-2-(hydroxymethyl)-1,3-thiazole

Precaución: for Research Use Only

Formula: C₄H₄BrNOS
MW: 194.05 g/mol
MDL Number: MFCD21728921
CAS Number: 911052-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR